Assay Details
| Gene Symbol: LOC100767953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503218.2 | XP_003503266.2 | 8 - 9 | 922 | 87 | |
| XM_003503219.2 | XP_003503267.1 | 8 - 9 | 1522 | 87 | ||
| XM_007643827.1 | XP_007642017.1 | 9 - 10 | 936 | 87 | ||
| XM_007643828.1 | XP_007642018.1 | 9 - 10 | 915 | 87 | ||
| XM_007643829.1 | XP_007642019.1 | 7 - 8 | 3381 | 87 | ||
More Information
Set Membership: |
|

LOC100767953
Genomic Map