Assay Details
| Gene Symbol: Gtf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652294.1 | XP_007650484.1 | 2 - 3 | 297 | 59 | |
| GenBank mRNA | JI865919.1 | - | 1 - 2 | 192 | 59 | |
More Information
Set Membership: |
|

Gtf3a
Genomic Map