Assay Details
| Gene Symbol: Lca5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513234.2 | XP_003513282.2 | 8 - 9 | 1571 | 67 | |
| XM_007653541.1 | XP_007651731.1 | 8 - 9 | 1504 | 67 | ||
More Information
Set Membership: |
|

Lca5l
Genomic Map