Assay Details
| Gene Symbol: Gfer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501544.2 | XP_003501592.1 | 2 - 3 | 251 | 72 | |
| GenBank mRNA | JP063138.1 | - | 1 - 2 | 162 | 72 | |
More Information
Set Membership: |
|

Gfer
Genomic Map