Assay Details
| Gene Symbol: Alg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502959.2 | XP_003503007.2 | 5 - 6 | 542 | 77 | |
| XM_007643553.1 | XP_007641743.1 | 5 - 6 | 623 | 77 | ||
| XM_007643554.1 | XP_007641744.1 | 4 - 5 | 577 | 77 | ||
| GenBank mRNA | JI867862.1 | - | 5 - 6 | 516 | 77 | |
More Information
Set Membership: |
|

Alg5
Genomic Map