Assay Details
| Gene Symbol: Alg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509608.2 | XP_003509656.1 | 8 - 9 | 1179 | 65 | |
| XM_007650227.1 | XP_007648417.1 | 7 - 8 | 1100 | 65 | ||
| XM_007650229.1 | XP_007648419.1 | 7 - 8 | 1033 | 65 | ||
| GenBank mRNA | JI869911.1 | - | 8 - 9 | 1173 | 65 | |
| JP048657.1 | - | 8 - 9 | 1173 | 65 | ||
More Information
Set Membership: |
|

Alg3
Genomic Map