Assay Details
| Gene Symbol: Mpst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514198.2 | XP_003514246.2 | 2 - 3 | 738 | 101 | |
| XM_007654377.1 | XP_007652567.1 | 2 - 3 | 813 | 101 | ||
| GenBank mRNA | JI890107.1 | - | 2 - 3 | 644 | 101 | |
| JP056780.1 | - | 2 - 3 | 644 | 101 | ||
More Information
Set Membership: |
|

Mpst
Genomic Map