Assay Details
| Gene Symbol: LOC100756710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515100.2 | XP_003515148.1 | 3 - 4 | 450 | 100 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100756710| Gene Symbol: LOC100756710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515100.2 | XP_003515148.1 | 3 - 4 | 450 | 100 | |