Assay Details
| Gene Symbol: LOC100756710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515100.2 | XP_003515148.1 | 4 - 5 | 532 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100756710| Gene Symbol: LOC100756710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515100.2 | XP_003515148.1 | 4 - 5 | 532 | 99 | |
Set Membership: |
|