Assay Details
| Gene Symbol: Adm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648890.1 | XP_007647080.1 | 2 - 3 | 302 | 65 | |
| GenBank mRNA | JI885869.1 | - | 2 - 3 | 284 | 65 | |
More Information
Set Membership: |
|

Adm
Genomic Map