Assay Details
| Gene Symbol: Sox30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643426.1 | XP_007641616.1 | 4 - 5 | 701 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sox30| Gene Symbol: Sox30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643426.1 | XP_007641616.1 | 4 - 5 | 701 | 66 | |
Set Membership: |
|