Assay Details
| Gene Symbol: Fkbp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501023.2 | XP_003501071.2 | 1 - 2 | 345 | 80 | |
| XM_007641736.1 | XP_007639926.1 | 1 - 2 | 345 | 80 | ||
| GenBank mRNA | JI866466.1 | - | 1 - 2 | 315 | 80 | |
| JP061535.1 | - | 1 - 2 | 315 | 80 | ||
More Information
Set Membership: |
|

Fkbp7
Genomic Map