Assay Details
| Gene Symbol: LOC100770307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI884603.1 | - | 3 - 4 | 322 | 107 | |
| JP038796.1 | - | 3 - 4 | 322 | 107 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100770307| Gene Symbol: LOC100770307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JI884603.1 | - | 3 - 4 | 322 | 107 | |
| JP038796.1 | - | 3 - 4 | 322 | 107 | ||
Set Membership: |
|