Assay Details
| Gene Symbol: LOC100770307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654535.1 | XP_007652725.1 | 3 - 4 | 541 | 70 | |
| XM_007654536.1 | XP_007652726.1 | 3 - 4 | 539 | 70 | ||
| XM_007654537.1 | XP_007652727.1 | 3 - 4 | 541 | 70 | ||
| XM_007654538.1 | XP_007652728.1 | 3 - 4 | 538 | 70 | ||
More Information
Set Membership: |
|

LOC100770307
Genomic Map