Assay Details
| Gene Symbol: Calu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497517.2 | XP_003497565.1 | 6 - 7 | 933 | 94 | |
| XM_007613410.1 | XP_007611600.1 | 6 - 7 | 933 | 94 | ||
| GenBank mRNA | JI896143.1 | - | 3 - 4 | 432 | 94 | |
| JP058926.1 | - | 3 - 4 | 431 | 94 | ||
More Information
Set Membership: |
|

Calu
Genomic Map