Assay Details
| Gene Symbol: Kcnj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505308.2 | XP_003505356.1 | 2 - 3 | 916 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnj3| Gene Symbol: Kcnj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505308.2 | XP_003505356.1 | 2 - 3 | 916 | 64 | |
Set Membership: |
|