Assay Details
| Gene Symbol: LOC100763541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512866.2 | XP_003512914.1 | 4 - 5 | 581 | 62 | |
| GenBank mRNA | JI889607.1 | - | 2 - 3 | 146 | 62 | |
| JP043874.1 | - | 2 - 3 | 146 | 62 | ||
More Information
Set Membership: |
|

LOC100763541
Genomic Map