Assay Details
| Gene Symbol: LOC100763541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512866.2 | XP_003512914.1 | 5 - 6 | 665 | 76 | |
| GenBank mRNA | JI889607.1 | - | 3 - 4 | 230 | 76 | |
| JP043874.1 | - | 3 - 4 | 230 | 76 | ||
More Information
Set Membership: |
|

LOC100763541
Genomic Map