Assay Details
| Gene Symbol: Ccdc53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499695.2 | XP_003499743.1 | 5 - 6 | 552 | 96 | |
| XM_007640424.1 | XP_007638614.1 | 5 - 6 | 532 | 96 | ||
| XM_007640425.1 | XP_007638615.1 | 5 - 6 | 552 | 96 | ||
| XM_007640426.1 | XP_007638616.1 | 5 - 6 | 552 | 96 | ||
| GenBank mRNA | JP038886.1 | - | 5 - 6 | 546 | 96 | |
More Information
Set Membership: |
|

Ccdc53
Genomic Map