Assay Details
| Gene Symbol: Zdhhc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505354.2 | XP_003505402.1 | 4 - 5 | 570 | 66 | |
| XM_007645888.1 | XP_007644078.1 | 4 - 5 | 570 | 66 | ||
| XM_007645889.1 | XP_007644079.1 | 3 - 4 | 285 | 66 | ||
| GenBank mRNA | JP040685.1 | - | 3 - 4 | 557 | 66 | |
More Information
Set Membership: |
|

Zdhhc3
Genomic Map