Assay Details
| Gene Symbol: Tnfrsf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506737.2 | XP_003506785.1 | 2 - 3 | 825 | 63 | |
| XM_007647272.1 | XP_007645462.1 | 3 - 4 | 900 | 63 | ||
| GenBank mRNA | JP040772.1 | - | 2 - 3 | 752 | 63 | |
More Information
Set Membership: |
|

Tnfrsf21
Genomic Map