Assay Details
| Gene Symbol: Mrpl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509973.2 | XP_003510021.1 | 2 - 3 | 249 | 65 | |
| GenBank mRNA | JI865232.1 | - | 2 - 3 | 231 | 65 | |
| JP062182.1 | - | 2 - 3 | 231 | 65 | ||
More Information
Set Membership: |
|

Mrpl20
Genomic Map