Assay Details
| Gene Symbol: Polr2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514298.2 | XP_003514346.2 | 3 - 4 | 419 | 66 | |
| GenBank mRNA | JP052590.1 | - | 1 - 2 | 83 | 66 | |
More Information
Set Membership: |
|

Polr2g
Genomic Map