Assay Details
| Gene Symbol: LOC100766665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607549.1 | XP_007605739.1 | 4 - 5 | 531 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100766665| Gene Symbol: LOC100766665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607549.1 | XP_007605739.1 | 4 - 5 | 531 | 76 | |
Set Membership: |
|