Assay Details
| Gene Symbol: Tac4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503964.2 | XP_003504012.2 | 3 - 4 | 995 | 65 | |
| XM_007644480.1 | XP_007642670.1 | 3 - 4 | 995 | 65 | ||
| XM_007644481.1 | XP_007642671.1 | 3 - 4 | 995 | 65 | ||
More Information
Set Membership: |
|

Tac4
Genomic Map