Assay Details
| Gene Symbol: LOC100764098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510592.1 | XP_003510640.1 | 5 - 6 | 772 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100764098| Gene Symbol: LOC100764098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510592.1 | XP_003510640.1 | 5 - 6 | 772 | 70 | |
Set Membership: |
|