Assay Details
| Gene Symbol: Slc25a30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641008.1 | XP_007639198.1 | 2 - 3 | 279 | 79 | |
| XM_007641009.1 | XP_007639199.1 | 2 - 3 | 279 | 79 | ||
More Information
Set Membership: |
|

Slc25a30
Genomic Map