Assay Details
| Gene Symbol: Ip6k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510823.2 | XP_003510871.1 | 3 - 4 | 667 | 55 | |
| XM_007651405.1 | XP_007649595.1 | 3 - 4 | 588 | 55 | ||
More Information
Set Membership: |
|

Ip6k3
Genomic Map