Assay Details
| Gene Symbol: Klf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648323.1 | XP_007646513.1 | 6 - 7 | 1660 | 60 | |
| XM_007648324.1 | XP_007646514.1 | 6 - 7 | 1140 | 60 | ||
| XM_007648325.1 | XP_007646515.1 | 5 - 6 | 1481 | 60 | ||
More Information
Set Membership: |
|

Klf8
Genomic Map