Assay Details
| Gene Symbol: Ssbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503024.2 | XP_003503072.1 | 4 - 5 | 355 | 65 | |
| GenBank mRNA | JI887809.1 | - | 2 - 3 | 203 | 65 | |
| JP049739.1 | - | 4 - 5 | 339 | 65 | ||
More Information
Set Membership: |
|

Ssbp1
Genomic Map