Assay Details
| Gene Symbol: LOC100751176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506952.1 | XP_003507000.1 | 4 - 5 | 685 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100751176| Gene Symbol: LOC100751176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506952.1 | XP_003507000.1 | 4 - 5 | 685 | 66 | |
Set Membership: |
|