Assay Details
| Gene Symbol: LOC103162716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646043.1 | XP_007644233.1 | 19 - 20 | 2839 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103162716| Gene Symbol: LOC103162716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646043.1 | XP_007644233.1 | 19 - 20 | 2839 | 95 | |
Set Membership: |
|