Assay Details
| Gene Symbol: Rnf145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502790.2 | XP_003502838.1 | 7 - 8 | 1299 | 60 | |
| GenBank mRNA | JI888528.1 | - | 7 - 8 | 989 | 60 | |
| JP060898.1 | - | 7 - 8 | 989 | 60 | ||
More Information
Set Membership: |
|

Rnf145
Genomic Map