Assay Details
| Gene Symbol: Tmem255b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511176.1 | XP_003511224.1 | 4 - 5 | 350 | 59 | |
| XM_003511177.1 | XP_003511225.1 | 3 - 4 | 287 | 59 | ||
More Information
Set Membership: |
|

Tmem255b
Genomic Map