Assay Details
| Gene Symbol: S100a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642300.1 | XP_007640490.1 | 2 - 3 | 355 | 100 | |
| GenBank mRNA | JP056514.1 | - | 2 - 3 | 301 | 100 | |
More Information
Set Membership: |
|

S100a13
Genomic Map