Assay Details
| Gene Symbol: LOC100752363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649819.1 | XP_007648009.1 | 1550 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100752363| Gene Symbol: LOC100752363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649819.1 | XP_007648009.1 | 1550 | 76 | ||