Assay Details
| Gene Symbol: LOC100752363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007649819.1 | XP_007648009.1 | 190 | 104 | ||
| GenBank mRNA | JP049521.1 | - | 2 - 3 | 190 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_007649819.1 transcripts.
Set Membership: |
|

LOC100752363
Genomic Map