Assay Details
| Gene Symbol: Kiaa0101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513592.2 | XP_003513640.1 | 1 - 2 | 178 | 61 | |
| GenBank mRNA | JI894327.1 | - | 1 - 2 | 183 | 61 | |
| JP041864.1 | - | 1 - 2 | 183 | 61 | ||
More Information
Set Membership: |
|

Kiaa0101
Genomic Map