Assay Details
| Gene Symbol: Mfrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514383.1 | XP_003514431.1 | 12 - 13 | 1531 | 62 | |
| XM_003514385.1 | XP_003514433.1 | 12 - 13 | 1513 | 62 | ||
More Information
Set Membership: |
|

Mfrp
Genomic Map