Assay Details
| Gene Symbol: Snx12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511049.2 | XP_003511097.2 | 3 - 4 | 538 | 72 | |
| GenBank mRNA | JI886535.1 | - | 3 - 4 | 502 | 72 | |
| JP047104.1 | - | 3 - 4 | 500 | 72 | ||
More Information
Set Membership: |
|

Snx12
Genomic Map