Assay Details
| Gene Symbol: Tbcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646052.1 | XP_007644242.1 | 1 - 2 | 240 | 63 | |
| GenBank mRNA | JI875273.1 | - | 1 - 2 | 240 | 63 | |
More Information
Set Membership: |
|

Tbcb
Genomic Map