Assay Details
| Gene Symbol: LOC100769148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512489.2 | XP_003512537.1 | 14 - 15 | 1484 | 85 | |
| GenBank mRNA | JI866047.1 | - | 12 - 13 | 1177 | 85 | |
| JP044874.1 | - | 12 - 13 | 1177 | 85 | ||
More Information
Set Membership: |
|

LOC100769148
Genomic Map