Assay Details
| Gene Symbol: LOC100769148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512489.2 | XP_003512537.1 | 3 - 4 | 405 | 132 | |
| GenBank mRNA | JI866047.1 | - | 1 - 2 | 98 | 132 | |
| JP044874.1 | - | 1 - 2 | 98 | 132 | ||
More Information
Set Membership: |
|

LOC100769148
Genomic Map