Assay Details
| Gene Symbol: LOC100769148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512489.2 | XP_003512537.1 | 6 - 7 | 613 | 113 | |
| GenBank mRNA | JI866047.1 | - | 4 - 5 | 306 | 113 | |
| JP044874.1 | - | 4 - 5 | 306 | 113 | ||
More Information
Set Membership: |
|

LOC100769148
Genomic Map