Assay Details
| Gene Symbol: Arhgap35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513716.2 | XP_003513764.1 | 4 - 5 | 4230 | 67 | |
| GenBank mRNA | JI886679.1 | - | 4 - 5 | 66 | ||
| JP045978.1 | - | 4 - 5 | 3983 | 67 | ||
More Information
Set Membership: |
|

Arhgap35
Genomic Map