Assay Details
| Gene Symbol: Mdfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650914.1 | XP_007649104.1 | 2 - 3 | 184 | 89 | |
| XM_007650916.1 | XP_007649106.1 | 2 - 3 | 410 | 89 | ||
| XM_007650921.1 | XP_007649111.1 | 2 - 3 | 184 | 89 | ||
More Information
Set Membership: |
|

Mdfic
Genomic Map