Assay Details
| Gene Symbol: Emp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652680.1 | XP_007650870.1 | 3 - 4 | 409 | 67 | |
| GenBank mRNA | JI889306.1 | - | 1 - 2 | 145 | 67 | |
More Information
Set Membership: |
|

Emp3
Genomic Map