Assay Details
| Gene Symbol: Pgbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646111.1 | XP_007644301.1 | 3 - 4 | 2876 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgbd1| Gene Symbol: Pgbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646111.1 | XP_007644301.1 | 3 - 4 | 2876 | 68 | |
Set Membership: |
|