Assay Details
| Gene Symbol: Ywhab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504570.2 | XP_003504618.2 | 5 - 6 | 699 | 69 | |
| GenBank mRNA | JP044964.1 | - | 4 - 5 | 694 | 69 | |
More Information
Set Membership: |
|

Ywhab
Genomic Map