Assay Details
| Gene Symbol: Rab32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510970.2 | XP_003511018.1 | 1 - 2 | 397 | 58 | |
| GenBank mRNA | JI894410.1 | - | 1 - 2 | 307 | 58 | |
| JP054596.1 | - | 1 - 2 | 307 | 58 | ||
More Information
Set Membership: |
|

Rab32
Genomic Map